Invitrogen
Manufacturer Code:MA520935
Catalog # PIMA520935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E5 |
Immunogen | ACTR1A (NP&lowbar005727 279 a.a. &bsim 375 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Actin-RPV; ACTR1A ARP1 ARP1 actin-related protein 1 homolog A centractin alpha (yeast) actin-RPV centractin alpha ARP1 yeast homolog A centrosome-associated actin homolog ARP1 (actin-related protein 1 yeast) homolog A (centractin alpha) CTRN1 FLJ52695 FLJ52800 FLJ55002 ARP1 actin-related protein 1 homolog A centractin alpha; Alpha-centractin; ARP1; Centractin; Centrosome-associated actin homolog
Gene Aliases: ACTR1A; ARP1; Arp1A; CTRN1
UniProt ID: (Human) P61163
Entrez Gene ID: (Human) 10121
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.