Novus Biologicals
Manufacturer Code:NBP153075
Catalog # NBP153075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ACTR1A(ARP1 actin-related protein 1 homolog A centractin alpha (yeast)) The peptide sequence was selected from the middle region of ACTR1A. Peptide sequence AIKERACYLSINPQKDETLETEKAQYYLPDGSTIEIGPSRFRAPE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-RPV; Actin-RPV alpha-centractin ARP1 (actin-related protein 1 yeast) homolog A (centractin alpha) ARP1 actin-related protein 1 homolog A centractin alpha (yeast) ARP1actin-RPV centractin Centrosome-associated actin homolog CTRN1 FLJ52695 FLJ52800 FLJ55002; Alpha-centractin; ARP1; Centractin; Centrosome-associated actin homolog
Gene Aliases: ACTR1A; ARP1; Arp1A; CTRN1
UniProt ID: (Human) P61163
Entrez Gene ID: (Human) 10121
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.