Invitrogen
Manufacturer Code:PA544739
Catalog # PIPA544739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ACTR10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin related protein 10 homolog; Actin-related protein 10; actin-related protein 10 homolog; actin-related protein 10-like protein; Actin-related protein 11; ARP10 actin related protein 10 homolog; ARP10 actin-related protein 10 homolog; hARP11
Gene Aliases: Act11; ACTR10; ACTR11; Arp10; ARP11; HARP11; zgc:55635
UniProt ID: (Human) Q9NZ32, (Bovine) Q3ZBD2, (Mouse) Q9QZB7
Entrez Gene ID: (Rabbit) 100338265, (Human) 55860, (Bovine) 511083, (Dog) 480337, (Horse) 100058966, (Pig) 100620619, (Mouse) 56444, (Rat) 299121, (Guinea pig) 100715149, (Zebrafish) 393139
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.