Abnova
Manufacturer Code:H00000081M01
Catalog # 89-016-033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D10 |
Immunogen | ACTN4 (NP_004915 592 a.a. ~ 701 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ACTN4.
For Research Use Only
Protein Aliases: ACTININ-4; Alpha-actinin-4; DKFZp686K23158; focal segmental glomerulosclerosis 1; FSGS; FSGS1; Non-muscle alpha-actinin 4
Gene Aliases: ACTININ-4; ACTN4; FSGS; FSGS1
UniProt ID: (Human) O43707
Entrez Gene ID: (Human) 81
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.