Proteintech
Manufacturer Code:666281IG150UL
Catalog # 50-173-7046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E12B8 |
Immunogen | ACTN4 Fusion Protein Ag13549 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACTININ 4 actinin alpha 4 ACTN4 Alpha actinin 4 F actin cross linking protein FSGS FSGS1 Non muscle alpha actinin 4; alpha actinin 4; Alpha-actinin-4; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; Non-muscle alpha-actinin 4
Gene Aliases: ACTININ-4; ACTN4; C77391; FSGS; FSGS1
UniProt ID: (Human) O43707, (Mouse) P57780, (Rat) Q9QXQ0
Entrez Gene ID: (Human) 81, (Mouse) 60595, (Rat) 63836
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.