Invitrogen
Manufacturer Code:PA578719
Catalog # PIPA578719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human ACTN3. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | At 2-8°C for one year. Protect from light. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actinin, alpha 3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-3; Alpha-actinin-3 Alpha-actinin skeletal muscle isoform 3 F-actin cross-linking protein ACTN3; F-actin cross-linking protein
Gene Aliases: ACTN3
UniProt ID: (Human) Q08043
Entrez Gene ID: (Human) 89
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.