Novus Biologicals
Manufacturer Code:NBP234529IR
Catalog # NBP234529IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F6 |
Immunogen | Synthetic peptide corresponding to aa1-24 of human ACTH |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACTH; ACTH adrenocorticotropic hormone adrenocorticotropin alpha-melanocyte-stimulating hormone alpha-MSH beta-endorphin beta-LPH beta-melanocyte-stimulating hormone beta-MSH CLIP corticotropin-like intermediary peptide corticotropin-lipotropin gamma-LPH gamma-MSH lipotropin beta lipotropin gamma LPH melanotropin alpha melanotropin beta melanotropin gamma met-enkephalin MSH NPP POC pro-ACTH-endorphin proopiomelanocortin pro-opiomelanocortin proopiomelanocortin preproprotein; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte-stimulating hormone; Alpha-MSH; Beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; Beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; Corticotropin-lipotropin; Gamma-LPH; Gamma-MSH; Lipotropin beta; Lipotropin gamma; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; Melanotropin beta; Melanotropin gamma; Met-enkephalin; NPP; opiomelanocortin prepropeptide; POMC; Potential peptide; pro-ACTH-endorphin; Pro-opiomelanocortin; proopiomelanocortin preproprotein
Gene Aliases: ACTH; CLIP; LPH; MSH; NPP; POC; POMC
UniProt ID: (Human) P01189
Entrez Gene ID: (Human) 5443
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.