Abnova
Manufacturer Code:MAB15136
Catalog # 89-035-344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | RM257 |
Immunogen | A synthetic peptide corresponding to N-terminus of human ACTC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ACTC; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Alpha-cardiac actin; CMD1R; CMH11
Gene Aliases: ACTC; ACTC1; ASD5; CMD1R; CMH11; LVNC4
UniProt ID: (Human) P68032
Entrez Gene ID: (Human) 70
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.