Proteintech
Manufacturer Code:661251IG150UL
Catalog # 50-173-6561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F2B9 |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACTC ACTC1 ACTC1 cardiac muscle 1 ACTC1 specific Actin cardiac muscle specific Actin alpha cardiac muscle 1 actin alpha cardiac muscle 1 Alpha cardiac actin CMD1R CMH11; actin alpha cardiac; actin alpha cardiac 1; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha, cardiac 1; alpha-actin cardiac; Alpha-cardiac actin; alphac-actin; B-actin; cardiac muscle alpha actin 1
Gene Aliases: ACTC; Actc-1; ACTC1; ASD5; CMD1R; CMH11; LVNC4
UniProt ID: (Human) P68032, (Mouse) P68033, (Rat) P68035
Entrez Gene ID: (Human) 70, (Pig) 100152267, (Mouse) 11464, (Rat) 29275
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.