Invitrogen
Manufacturer Code:PA5116567
Catalog # PIPA5116567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-280 of human ACSS3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8430416H19Rik Acetate--CoA ligase 3 ACSS3 acyl-CoA synthetase short-chain family member 3 Acyl-CoA synthetase short-chain family member 3 mitochondrial AMP-binding enzyme 33217 Gm874 Propionate--CoA ligase RGD1307051; Acetate--CoA ligase 3; Acyl-CoA synthetase short-chain family member 3; Acyl-CoA synthetase short-chain family member 3, mitochondrial; AMP-binding enzyme, 33217; Propionate--CoA ligase
Gene Aliases: 8430416H19Rik; ACSS3; Gm874; RGD1307051
UniProt ID: (Human) Q9H6R3, (Mouse) Q14DH7, (Rat) A0A0G2K047
Entrez Gene ID: (Human) 79611, (Mouse) 380660, (Rat) 314800
Molecular Function:
dehydrogenase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.