Invitrogen
Manufacturer Code:PA526612
Catalog # PIPA526612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 31-60 amino acids from the N-terminal region of human ACSS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAS2 ACECS ACS ACSA dJ1161H23.1 acetate thiokinase acetate-CoA ligase acetyl-Coenzyme A synthetase 2 (ADP forming) acetyl-coenzyme A synthetase cytoplasmic acyl-activating enzyme cytoplasmic acetyl-coenzyme A synthetase; aceCS; AceCS1; acetate thiokinase; Acetate--CoA ligase; acetate-CoA ligase; Acetyl-CoA synthetase; Acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); Acetyl-coenzyme A synthetase, cytoplasmic; ACS; Acyl-activating enzyme; Acyl-CoA synthetase short-chain family member 2; cytoplasmic acetyl-coenzyme A synthetase; Propionate--CoA ligase
Gene Aliases: 1110017C11Rik; ACAS; Acas1; ACAS2; ACECS; AceCS1; ACS; Acs1; ACSA; ACSS2; dJ1161H23.1
UniProt ID: (Human) Q9NR19, (Mouse) Q9QXG4
Entrez Gene ID: (Human) 55902, (Mouse) 60525
Molecular Function: dehydrogenase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.