Proteintech
Manufacturer Code:171381AP150UL
Catalog # 50-173-1399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACSS1 Fusion Protein Ag10896 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAS2L AceCS2 AceCS2L Acetate CoA ligase 2 Acetyl CoA synthetase 2 ACSS1 KIAA1846; AceCS2; Acetate--CoA ligase 2; Acetyl-CoA synthetase 2; Acetyl-coenzyme A synthetase 2-like, mitochondrial; Acyl-CoA synthetase short-chain family member 1; Propionate--CoA ligase
Gene Aliases: 1110032O15Rik; Acas2; ACAS2L; ACECS1; AceCS2; AceCS2L; ACSS1; AI788978; KIAA1846
UniProt ID: (Human) Q5TF42, (Mouse) Q99NB1
Entrez Gene ID: (Human) 84532, (Mouse) 68738, (Rat) 296259
Molecular Function: dehydrogenase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.