Invitrogen
Manufacturer Code:PA560768
Catalog # PIPA560768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ACSM6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA synthetase medium-chain family member 6; Acyl-coenzyme A synthetase ACSM6 mitochondrial Acyl-CoA synthetase medium-chain family member 6 ACSM6 C10orf129; Acyl-coenzyme A synthetase ACSM6, mitochondrial; AMP-binding enzyme
Gene Aliases: ACSM6; bA310E22.3; C10orf129
UniProt ID: (Human) Q6P461
Entrez Gene ID: (Human) 142827
Molecular Function:
dehydrogenase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.