Novus Biologicals
Manufacturer Code:NBP271978AF750
Catalog # NP271978A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G1 |
Immunogen | Full length recombinant protein of human ACSM5 (NP_060358) produced in HEK293T cells. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Acyl-CoA synthetase medium-chain family member 5; acyl-CoA synthetase medium-chain family member 5 acyl-coenzyme A synthetase ACSM5 mitochondrial EC 6.2.1 EC 6.2.1.2 FLJ20581 MACS3; Acyl-coenzyme A synthetase ACSM5, mitochondrial
Gene Aliases: ACSM5; MACS3
UniProt ID: (Human) Q6NUN0
Entrez Gene ID: (Human) 54988
Molecular Function: dehydrogenase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.