Proteintech
Manufacturer Code:165911AP150UL
Catalog # 50-173-1040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACSM5 Fusion Protein Ag9373 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACSM5 MACS3; Acyl-CoA synthetase medium-chain family member 5; Acyl-coenzyme A synthetase ACSM5, mitochondrial
Gene Aliases: Aa2-174; ACSM5; C730019D22; C730027J19Rik; Cc1-38; MACS3; RGD1309578
UniProt ID: (Human) Q6NUN0, (Rat) Q6AYT9, (Mouse) Q8BGA8
Entrez Gene ID: (Human) 54988, (Rat) 361637, (Mouse) 272428
Molecular Function: dehydrogenase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.