Invitrogen
Manufacturer Code:PA5100374
Catalog # PIPA5100374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACSM3 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acsm3 Acyl-CoA synthetase medium-chain family member 3 acyl-coenzyme A synthetase ACSM3 mitochondrial Butyrate--CoA ligase 3 butyryl coenzyme A synthetase 3 Butyryl-coenzyme A synthetase 3 middle-chain acyl-CoA synthetase 3 Propionate--CoA ligase Protein SA protein SA homolog Sa SA (rat hypertension-associated) homolog SA hypertension-associated homolog SA rat hypertension-associated homolog SA rat hypertension-associated protein Sah ssubA; Acyl-CoA synthetase medium-chain family member 3; Acyl-coenzyme A synthetase ACSM3, mitochondrial; Butyrate--CoA ligase 3; butyryl coenzyme A synthetase 3; Butyryl-coenzyme A synthetase 3; Middle-chain acyl-CoA synthetase 3; Propionate--CoA ligase; Protein SA; Protein SA homolog; SA (rat hypertension-associated) homolog; SA hypertension-associated homolog; SA rat hypertension-associated homolog; SA rat hypertension-associated protein; ssubA
Gene Aliases: ACSM3; SA; SAH
UniProt ID: (Human) Q53FZ2, (Mouse) Q3UNX5, (Rat) Q6SKG1
Entrez Gene ID: (Human) 6296, (Mouse) 20216, (Rat) 24763
Molecular Function: dehydrogenase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.