Invitrogen
Manufacturer Code:PA561208
Catalog # PIPA561208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ACSM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA synthetase medium-chain family member 1; Acyl-coenzyme A synthetase ACSM1, mitochondrial; Benzoate--CoA ligase; BUCS1 MACS1 Butyrate CoA ligase acyl-coenzyme A synthetase ACSM1 mitochondrial butyrate--CoA ligase 1 butyryl Coenzyme A synthetase 1 butyryl-coenzyme A synthetase 1 lipoate-activating enzyme medium-chain acyl-CoA synthetase middle-chain acyl-CoA synthetase 1; Butyrate CoA ligase; Butyrate--CoA ligase 1; butyryl Coenzyme A synthetase 1; Butyryl-coenzyme A synthetase 1; Lipoate-activating enzyme; medium-chain acyl-CoA synthetase; Middle-chain acyl-CoA synthetase 1; Xenobiotic/medium-chain fatty acid-CoA ligase HXM-B
Gene Aliases: ACSM1; BUCS1; LAE; MACS1
UniProt ID: (Human) Q08AH1
Entrez Gene ID: (Human) 116285
Molecular Function:
dehydrogenase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.