Invitrogen
Manufacturer Code:PA559191
Catalog # PIPA559191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ACSL6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACS2 FACL6 LACS 6 LACS2 LACS5 fatty-acid-Coenzyme A ligase long-chain 6 long fatty acyl-CoA synthetase 2 long-chain-fatty-acid--CoA ligase 6; Arachidonate--CoA ligase; fatty acid Coenzyme A ligase, long chain 6; fatty-acid-Coenzyme A ligase, long-chain 6; LACS 6; long fatty acyl-CoA synthetase 2; Long-chain acyl-CoA synthetase 6; Long-chain-fatty-acid--CoA ligase 6; Long-chain-fatty-acid--CoA ligase, brain isozyme
Gene Aliases: A330035H04Rik; ACS2; ACSL6; AW050338; FACL6; KIAA0837; LACS; LACS 6; LACS2; LACS5; Lacsl; mKIAA0837
UniProt ID: (Human) Q9UKU0, (Mouse) Q91WC3, (Rat) P33124
Entrez Gene ID: (Human) 23305, (Mouse) 216739, (Rat) 117243
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.