Novus Biologicals
Manufacturer Code:NBP189269
Catalog # NBP189269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RYIINTADISTVIVDKPQKAVLLLEHVERKETPGLKLIILMDPFEEALKERGQKCGVVIKSMQAVEDCGQENHQAPVPPQPDDLSIVCFTSGTTGNPKGAMLTHGNVVADF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACS2long-chain 6 acyl-CoA synthetase long-chain family member 6 EC 6.2.1 EC 6.2.1.3 FLJ16173 LACS 6 long fatty acyl-CoA synthetase 2 Long-chain acyl-CoA synthetase 6 long-chain-fatty-acid--CoA ligase 6; Arachidonate--CoA ligase; fatty-acid-Coenzyme A ligase, long-chain 6; LACS 6; long fatty acyl-CoA synthetase 2; Long-chain acyl-CoA synthetase 6; Long-chain-fatty-acid--CoA ligase 6
Gene Aliases: ACS2; ACSL6; FACL6; KIAA0837; LACS 6; LACS2; LACS5
UniProt ID: (Human) Q9UKU0
Entrez Gene ID: (Human) 23305
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.