Invitrogen
Manufacturer Code:PA5114816
Catalog # PIPA5114816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACSL5 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700030F05Rik ACS2 Acs5 ACSL5 acyl-CoA synthetase long-chain family member 5 Arachidonate--CoA ligase Facl5 FACL5 for fatty acid coenzyme A ligase 5 fatty acid coenzyme A ligase 5 fatty acid Coenzyme A ligase long chain 5 fatty-acid-Coenzyme A ligase long-chain 5 HGNC:16526 LACS 5 long-chain acyl-CoA synthetase 5 long-chain fatty acid coenzyme A ligase 5 long-chain-fatty-acid--CoA ligase 5 UNQ633/PRO1250; Arachidonate--CoA ligase; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty acid Coenzyme A ligase, long chain 5; fatty-acid-Coenzyme A ligase, long-chain 5; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; Long-chain-fatty-acid--CoA ligase 5
Gene Aliases: 1700030F05Rik; ACS2; ACS5; ACSL5; FACL5; UNQ633/PRO1250
UniProt ID: (Human) Q9ULC5, (Rat) O88813, (Mouse) Q8JZR0
Entrez Gene ID: (Human) 51703, (Rat) 94340, (Mouse) 433256
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.