Novus Biologicals
Manufacturer Code:NBP297725G
Catalog # NB047101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ACSL5 fragment. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACS2 ACS5FACL5 for fatty acid coenzyme A ligase 5 acyl-CoA synthetase long-chain family member 5 EC 6.2.1 EC 6.2.1.3 FACL5fatty acid coenzyme A ligase 5 fatty-acid-Coenzyme A ligase long-chain 5 LACS 5 Long-chain acyl-CoA synthetase 5 long-chain fatty acid coenzyme A ligase 5 long-chain-fatty-acid--CoA ligase 5; Arachidonate--CoA ligase; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty-acid-Coenzyme A ligase, long-chain 5; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; Long-chain-fatty-acid--CoA ligase 5
Gene Aliases: ACS2; ACS5; ACSL5; FACL5; UNQ633/PRO1250
UniProt ID: (Human) Q9ULC5
Entrez Gene ID: (Human) 51703
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.