Invitrogen
Manufacturer Code:PA5100033
Catalog # PIPA5100033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACSL4/FACL4 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430020A05Rik ACS4 ACSL4 acyl-CoA synthetase 4 acyl-CoA synthetase long-chain family member 4 Arachidonate--CoA ligase AU018108 FACL4 fatty acid Coenzyme A ligase long chain 4 fatty acid-Coenzyme A ligase long chain 4 fatty acid-Coenzyme A ligase long chain 4 fatty-acid-Coenzyme A ligase long-chain 4 L MRX63 LACS 4 LACS4 lignoceroyl-CoA synthase long-chain acyl-CoA synthetase 4 long-chain fatty-acid-Coenzyme A ligase 4 long-chain-fatty-acid--CoA ligase 4 mACS4 MRX63 MRX68; acyl-CoA synthetase 4; Arachidonate--CoA ligase; fatty acid-Coenzyme A ligase, long chain 4; fatty-acid-Coenzyme A ligase, long-chain 4; LACS 4; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; Long-chain-fatty-acid--CoA ligase 4; mACS4
Gene Aliases: 9430020A05Rik; ACS4; ACSL4; AU018108; FACL4; LACS4; MRX63; MRX68
UniProt ID: (Human) O60488, (Mouse) Q9QUJ7
Entrez Gene ID: (Human) 2182, (Mouse) 50790
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.