Abnova
Manufacturer Code:PAB2504
Catalog # 89-126-716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 244 -274 at internal region of human ACSL4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ACSL4.
For Research Use Only
Protein Aliases: ACS4; acyl-CoA synthetase 4; Arachidonate--CoA ligase; FACL4; fatty-acid-Coenzyme A ligase, long-chain 4; LACS 4; LACS4; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; Long-chain-fatty-acid--CoA ligase 4; MRX63; MRX68
Gene Aliases: ACS4; ACSL4; FACL4; LACS4; MRX63; MRX68
UniProt ID: (Human) O60488
Entrez Gene ID: (Human) 2182
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.