Invitrogen
Manufacturer Code:PA5114815
Catalog # PIPA5114815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACSL3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610510B12Rik Acs3 Acsl3 acyl-CoA synthetase long-chain family member 3 Arachidonate--CoA ligase Brain acyl-CoA synthetase II C85929 FACL3 fatty acid Coenzyme A ligase long chain 3 fatty-acid-Coenzyme A ligase long-chain 3 LACS 3 LACS3 lignoceroyl-CoA synthase long-chain acyl-CoA synthetase 3 Long-chain-fatty-acid--CoA ligase 3 Pro2194; Arachidonate--CoA ligase; Brain acyl-CoA synthetase II; fatty acid Coenzyme A ligase, long chain 3; fatty-acid-Coenzyme A ligase, long-chain 3; LACS 3; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 3; Long-chain-fatty-acid--CoA ligase 3
Gene Aliases: 2610510B12Rik; ACS3; ACSL3; C85929; FACL3; LACS3; PRO2194
UniProt ID: (Human) O95573, (Rat) Q63151, (Mouse) Q9CZW4
Entrez Gene ID: (Human) 2181, (Rat) 114024, (Mouse) 74205
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.