Invitrogen
Manufacturer Code:PA517136
Catalog # PIPA517136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a sequence of human ACSL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACS1; Acyl CoA synthetase, long chain; Acyl-CoA synthetase 1; Arachidonate--CoA ligase; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; LACS 1; LACS 1 LACS 2 OTTHUMP00000218457 OTTHUMP00000218460 OTTHUMP00000218462 OTTHUMP00000218465 acyl-CoA synthetase 1 fatty-acid-Coenzyme A ligase long-chain 1 fatty-acid-Coenzyme A ligase long-chain 2 lignoceroyl-CoA synthase long-chain acyl-CoA synthetase 1 long-chain acyl-CoA synthetase 2 long-chain fatty acid-CoA ligase 2 long-chain fatty-acid-coenzyme A ligase 1 long-chain-fatty-acid--CoA ligase 1 palmitoyl-CoA ligase 1 palmitoyl-CoA ligase 2 paltimoyl-CoA ligase 1; LACS 2; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; Long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; Long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; Palmitoyl-CoA ligase 1; Palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase
Gene Aliases: Acas; Acas1; ACS; ACS1; ACSL1; Acsl2; COAA; FACL1; FACL2; FACS; LACS; LACS 1; LACS1; LACS2
UniProt ID: (Human) P33121, (Mouse) P41216, (Rat) P18163
Entrez Gene ID: (Human) 2180, (Mouse) 14081, (Rat) 25288
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.