Novus Biologicals
Manufacturer Code:NBP215251
Catalog # NBP215251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ACSL1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: ACS1; ACS1LACS 2 Acyl-CoA synthetase 1 acyl-CoA synthetase long-chain family member 1 EC 6.2.1 FACL1EC 6.2.1.3 fatty-acid-Coenzyme A ligase long-chain 1 LACS 1 LACSlong-chain 2 lignoceroyl-CoA synthase Long-chain acyl-CoA synthetase 1 Long-chain acyl-CoA synthetase 2 Long-chain fatty acid-CoA ligase 2 long-chain fatty-acid-coenzyme A ligase 1 long-chain-fatty-acid--CoA ligase 1 Palmitoyl-CoA ligase 1 Palmitoyl-CoA ligase 2 paltimoyl-CoA ligase 1; Acyl-CoA synthetase 1; Arachidonate--CoA ligase; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; LACS 1; LACS 2; lignoceroyl-CoA synthase; Long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; Long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; Long-chain-fatty-acid--CoA ligase 1; Palmitoyl-CoA ligase 1; Palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase
Gene Aliases: ACS1; ACSL1; FACL1; FACL2; LACS; LACS1; LACS2
UniProt ID: (Human) P33121
Entrez Gene ID: (Human) 2180
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.