Novus Biologicals
Manufacturer Code:NBP2839240.1ML
Catalog # NB031853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human ACSF3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA synthetase family member 3; acyl-CoA synthetase family member 3 acyl-CoA synthetase family member 3 mitochondrial EC 6.2.1 EC 6.2.1.- FLJ39242; Malonate--CoA ligase ACSF3, mitochondrial; malonyl-CoA synthetase
Gene Aliases: ACSF3; PSEC0197
UniProt ID: (Human) Q4G176
Entrez Gene ID: (Human) 197322
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.