Novus Biologicals
Manufacturer Code:NBP272220APC
Catalog # NB272220APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6E4 |
Immunogen | Full length recombinant protein of human ACSF2 (NP_079425) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ACSMW acyl-CoA synthetase family member 2 AVYV493 EC 6.2.1 EC 6.2.1.- EC 6.2.1.26 mitochondrial PPARG binding long chain fatty acid acyl Co-A ligase like; Medium-chain acyl-CoA ligase ACSF2, mitochondrial; PPARG binding, long chain fatty acid acyl Co-A ligase like
Gene Aliases: ACSF2; ACSMW; AVYV493; UNQ493/PRO1009
UniProt ID: (Human) Q96CM8
Entrez Gene ID: (Human) 80221
Molecular Function:
dehydrogenase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.