Invitrogen
Manufacturer Code:PA526853
Catalog # PIPA526853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 237-265 amino acids from the central region of human ACBG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA synthetase bubblegum family member 2; Arachidonate--CoA ligase ACSBG2; BGR BRGL PRTD-NY3 PRTDNY3 bubblegum-related protein long-chain-fatty-acid--CoA ligase ACSBG2; Bubblegum-related protein; Long-chain-fatty-acid--CoA ligase ACSBG2; PRTD-NY3; testicular tissue protein Li 8
Gene Aliases: ACSBG2; BGR; BRGL; PRTD-NY3; PRTDNY3; UNQ2443/PRO5005
UniProt ID: (Human) Q5FVE4, (Mouse) Q2XU92
Entrez Gene ID: (Human) 81616, (Mouse) 328845
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.