Proteintech
Manufacturer Code:160771AP150UL
Catalog # 50-173-0695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACSBG1 Fusion Protein Ag9043 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACSBG1 BG1 BGM GR LACS hBG1 hsBG hsBGM KIAA0631 Lipodisin LPD; Acyl-CoA synthetase bubblegum family member 1; bubblegum; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; GR-LACS; hBG1; Lipidosin; lipidosis-related protein lipidosin; lipodisin; Long-chain-fatty-acid--CoA ligase ACSBG1; mBG1; very long-chain acyl-CoA synthetase
Gene Aliases: ACSBG1; BG; BG1; BGM; E230019G03Rik; GR-LACS; Grlacs; KIAA0631; LPD; mKIAA0631; R75185
UniProt ID: (Human) Q96GR2, (Mouse) Q99PU5, (Rat) Q924N5
Entrez Gene ID: (Human) 23205, (Mouse) 94180, (Rat) 171410
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.