Abnova
Manufacturer Code:MAB3824
Catalog # 89-111-615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Hs-14 |
Immunogen | Native purified human VCP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Mouse monoclonal antibody raised against native purified ACRV1.
For Research Use Only
Protein Aliases: 15S Mg(2+)-ATPase p97 subunit; epididymis luminal protein 220; epididymis secretory protein Li 70; IBMPFD; MGC131997; MGC148092; MGC8560; p97; TER ATPase; TERA; Transitional endoplasmic reticulum ATPase; Valosin-containing protein; VCP; yeast Cdc48p homolog
Gene Aliases: ALS14; CMT2Y; HEL-220; HEL-S-70; IBMPFD; IBMPFD1; p97; TERA; VCP
UniProt ID: (Human) P55072
Entrez Gene ID: (Human) 7415
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.