Invitrogen
Manufacturer Code:PA571954
Catalog # PIPA571954
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 599-628 amino acids from the C-terminal region of human ACRC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acidic repeat-containing protein; Acidic repeat-containing protein ACRC; Germ cell nuclear antigen; putative nuclear protein
Gene Aliases: ACRC; GCNA; NAAR1
UniProt ID: (Human) Q96QF7
Entrez Gene ID: (Human) 93953
Molecular Function:
HMG box transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.