Invitrogen
Manufacturer Code:PA5101205
Catalog # PIPA5101205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACRBP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACRBP; Acrbp acrbp {ECO:0000312 acrosin binding protein acrosin-binding protein Cancer/testis antigen 23 Cancer/testis antigen OY-TES-1 CT23 EMBL:AAH79212.1} OY-TES-1 proacrosin binding protein proacrosin binding protein sp32 proacrosin-binding protein sp32 SP32 testicular tissue protein Li 10; acrbp {ECO:0000312|EMBL:AAH79212.1}; Acrosin-binding protein; Acrosin-binding protein, 32 kDa form, mature form; Acrosin-binding protein, 60 kDa form; Acrosin-binding protein, mature form; Cancer/testis antigen 23; Cancer/testis antigen OY-TES-1; CT23; proacrosin binding protein; proacrosin binding protein sp32; Proacrosin-binding protein sp32; testicular tissue protein Li 10
Gene Aliases: ACRBP; CT23; OY-TES-1; SP32
UniProt ID: (Human) Q8NEB7, (Rat) Q6AY33
Entrez Gene ID: (Human) 84519, (Rat) 500316
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.