Novus Biologicals
Manufacturer Code:NBP16244020UL
Catalog # NBP16244020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ACPT(acid phosphatase testicular) The peptide sequence was selected from the middle region of ACPT (NP_149059). Peptide sequence TLLALQGALGLYDGHTPPYAACLGFEFRKHLGNPAKDGGNVTVSLFYRND. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid phosphatase 4; acid phosphatase testicular EC 3.1.3.2 testicular acid phosphatase; Testicular acid phosphatase
Gene Aliases: ACP4; ACPT
UniProt ID: (Human) Q9BZG2
Entrez Gene ID: (Human) 93650
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.