Proteintech
Manufacturer Code:244101AP150UL
Catalog # 50-173-4339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACPP Fusion Protein Ag19701 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-nucleotidase; Acid phosphatase 3; acid phosphatase prostate ACP 3 ACP3 ACPP PAP Prostatic acid phosphatase; Ecto-5'-nucleotidase; PAP; PAPf39; Prostatic acid phosphatase; prostatic acid phosphotase; Protein tyrosine phosphatase ACP3; Thiamine monophosphatase; TMPase
Gene Aliases: 5'-NT; ACP-3; ACP3; ACPP
UniProt ID: (Human) P15309
Entrez Gene ID: (Human) 55
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.