Invitrogen
Manufacturer Code:PA572305
Catalog # PIPA572305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 116-146 amino acids from of human ACP6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid phosphatase 6, lysophosphatidic; acid phosphatase like 1; Acid phosphatase-like protein 1; Lysophosphatidic acid phosphatase type 6; Lysophosphatidic acid phosphatase type 6 3.1.3.2 Acid phosphatase 6 lysophosphatidic Acid phosphatase-like protein 1 PACPL1 ACP6 ACPL1 LPAP; PACPL1
Gene Aliases: 5730559A09Rik; Acp6; ACPL1; AU022842; Lpap; mPACPL1
UniProt ID: (Mouse) Q3TNE2
Entrez Gene ID: (Mouse) 66659
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.