Novus Biologicals
Manufacturer Code:NBP299166UV
Catalog # NB058583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human ACP6. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase 6 lysophosphatidicPACPL1 Acid phosphatase-like protein 1 ACPL1acid phosphatase like 1 EC 3.1.3.2 LPAPlysophosphatidic acid phosphatase type 6; Acid phosphatase 6, lysophosphatidic; Acid phosphatase-like protein 1; Lysophosphatidic acid phosphatase type 6; PACPL1
Gene Aliases: ACP6; ACPL1; LPAP; PACPL1; UNQ205/PRO231
UniProt ID: (Human) Q9NPH0
Entrez Gene ID: (Human) 51205
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.