Invitrogen
Manufacturer Code:PA5106914
Catalog # PIPA5106914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACP5 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase 5 tartrate resistant acid phosphatase type V ACP5 HPAP human purple acid phosphatase T5ap Tartrate-resistant acid ATPase tartrate-resistant acid phosphatase 5a tartrate-resistant acid phosphatase 5b tartrate-resistant acid phosphatase type 5 TRACP TRACP5a TRACP5b Trap TR-AP TRAPase TrATPase TTRRAP type 5 acid phosphatase; acid phosphatase type V; human purple acid phosphatase; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; Tartrate-resistant acid phosphatase type 5; TR-AP; TrATPase; Type 5 acid phosphatase
Gene Aliases: ACP5; HPAP; T5ap; TRACP; TRACP5a; TRACP5b; TRAP; TrATPase; TTRRAP
UniProt ID: (Human) P13686, (Rat) P29288, (Mouse) Q05117
Entrez Gene ID: (Human) 54, (Rat) 25732, (Mouse) 11433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.