Novus Biologicals
Manufacturer Code:NBP16249120UL
Catalog # NBP1624920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ACP2(acid phosphatase 2 lysosomal) The peptide sequence was selected from the middle region of ACP2 (NP_001601). Peptide sequence VPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANET. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase 2 lysosomal EC 3.1.3.2 LAP lysosomal acid phosphatase; LAP; Lysosomal acid phosphatase
Gene Aliases: ACP2; LAP
UniProt ID: (Human) P11117
Entrez Gene ID: (Human) 53
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.