Proteintech
Manufacturer Code:152361AP150UL
Catalog # 50-173-0085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACP2 Fusion Protein Ag7356 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase 2 lysosomal ACP2 LAP Lysosomal acid phosphatase; Acid phosphatase 2, lysozymal; LAP; LMW-PTP-II low molecular weight phosphotyrosine protein phosphatase isozyme 2; Lysosomal acid phosphatase
Gene Aliases: Acp-2; ACP2; LAP
UniProt ID: (Human) P11117, (Mouse) P24638
Entrez Gene ID: (Human) 53, (Rat) 24162, (Mouse) 11432
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.