Invitrogen
Manufacturer Code:PA127656
Catalog # PIPA127656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human Protein-tyrosine-phosphatase. isoform 3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase of erythrocyte; Adipocyte acid phosphatase; cytoplasmic phosphotyrosyl protein phosphatase; HAAP MGC111030 MGC3499; LMW-PTP; LMW-PTPase; Low molecular weight cytosolic acid phosphatase; Low molecular weight phosphotyrosine protein phosphatase; protein tyrosine phosphatase; Red cell acid phosphatase 1; testicular secretory protein Li 37
Gene Aliases: ACP1; HAAP; LMW-PTP
UniProt ID: (Human) P24666
Entrez Gene ID: (Human) 52
Molecular Function:
hydrolase
oxidoreductase
phosphatase
protein phosphatase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.