Invitrogen
Manufacturer Code:PA570858
Catalog # PIPA570858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human ACOXL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA oxidase-like protein; acyl-CoA oxidase-like protein acyl-coenzyme A oxidase-like protein; acyl-Coenzyme A oxidase-like; Acyl-coenzyme A oxidase-like protein; acyl-coenzyme A oxidase-like protein-like
Gene Aliases: ACOXL
UniProt ID: (Human) Q9NUZ1
Entrez Gene ID: (Human) 55289, (Dog) 611729, (Pig) 100520554, (Horse) 100064086, (Rabbit) 108176402
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.