Novus Biologicals
Manufacturer Code:NBP298600AF532
Catalog # NB054430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ACOX3 fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acyl-CoA oxidase 3 pristanoyl Branched-chain acyl-CoA oxidase BRCACox BRCOX EC 1.3.3.6 peroxisomal acyl-coenzyme A oxidase 3 PRCOX pristanoyl Pristanoyl-CoA oxidase; acyl-Coenzyme A oxidase 3, pristanoyl; Branched-chain acyl-CoA oxidase; BRCACox; Peroxisomal acyl-coenzyme A oxidase 3; Pristanoyl-CoA oxidase
Gene Aliases: ACOX3; BRCOX; PRCOX
UniProt ID: (Human) O15254
Entrez Gene ID: (Human) 8310
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.