Invitrogen
Manufacturer Code:PA5114814
Catalog # PIPA5114814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACOX2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase; 3-alpha7-alpha12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase 3-alpha7-alpha12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase ACOX2 acyl-CoA oxidase 2 acyl-CoA oxidase 2 branched chain acyl-Coenzyme A oxidase 2 branched chain BCOX branched chain acyl-CoA oxidase BRCACOX BRCOX peroxisomal acyl-coenzyme A oxidase 2 peroxisomal branched chain acyl-CoA oxidase THCA-CoA oxidase THCCox trihydroxycoprostanoyl-CoA oxidase; acyl-CoA oxidase 2, branched chain; acyl-Coenzyme A oxidase 2, branched chain; branched chain acyl-CoA oxidase; Peroxisomal acyl-coenzyme A oxidase 2; peroxisomal branched chain acyl-CoA oxidase; THCA-CoA oxidase; Trihydroxycoprostanoyl-CoA oxidase
Gene Aliases: ACOX2; BCOX; BRCACOX; BRCOX; THCCox
UniProt ID: (Human) Q99424, (Mouse) Q9QXD1
Entrez Gene ID: (Human) 8309, (Rat) 252898, (Mouse) 93732
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.