Novus Biologicals
Manufacturer Code:NB10057853
Catalog # NB10057853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QWAQKSPTNTQENP corresponding to internal region (near C-Terminus) according to NP_003491.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase 12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase 3-alpha 7-alpha acyl-CoA oxidase 2 branched chain acyl-Coenzyme A oxidase 2 branched chain BCOX BRCACOXTHCA-CoA oxidase BRCOX EC 1.17.99.3 peroxisomal acyl-coenzyme A oxidase 23-alpha peroxisomal branched chain acyl-CoA oxidase THCCox Trihydroxycoprostanoyl-CoA oxidase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase; acyl-CoA oxidase 2, branched chain; acyl-Coenzyme A oxidase 2, branched chain; branched chain acyl-CoA oxidase; Peroxisomal acyl-coenzyme A oxidase 2; peroxisomal branched chain acyl-CoA oxidase; THCA-CoA oxidase; Trihydroxycoprostanoyl-CoA oxidase
Gene Aliases: ACOX2; BCOX; BRCACOX; BRCOX; THCCox
UniProt ID: (Human) Q99424
Entrez Gene ID: (Human) 8309
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.