Invitrogen
Manufacturer Code:MA530031
Catalog # PIMA530031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 102 |
Immunogen | Recombit Human ACOX1/AOX protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: acyl-CoA oxidase 1, palmitoyl; acyl-CoA oxidase, straight-chain; Acyl-coenzyme A oxidase 1 peroxisomal Palmitoyl-CoA oxidase AOX; acyl-Coenzyme A oxidase 1, palmitoyl; AOX; Palmitoyl-CoA oxidase; Peroxisomal acyl-CoA oxidase 1, A chain; Peroxisomal acyl-CoA oxidase 1, B chain; Peroxisomal acyl-CoA oxidase 1, C chain; Peroxisomal acyl-coenzyme A oxidase 1; peroxisomal fatty acyl-CoA oxidase; SCOX; Straight-chain acyl-CoA oxidase
Gene Aliases: ACOX; ACOX1; PALMCOX; SCOX
UniProt ID: (Human) Q15067
Entrez Gene ID: (Human) 51
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.