Novus Biologicals
Manufacturer Code:NBP215249
Catalog # NBP215249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ACOX1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACOXEC 1.3.3.6 acyl-CoA oxidase 1 palmitoyl acyl-CoA oxidase straight-chain acyl-Coenzyme A oxidase 1 palmitoyl AOX PALMCOX Palmitoyl-CoA oxidase peroxisomal acyl-coenzyme A oxidase 1 peroxisomal fatty acyl-CoA oxidase SCOXMGC1198 Straight-chain acyl-CoA oxidase; acyl-CoA oxidase 1, palmitoyl; acyl-CoA oxidase, straight-chain; acyl-Coenzyme A oxidase 1, palmitoyl; AOX; Palmitoyl-CoA oxidase; Peroxisomal acyl-CoA oxidase 1, A chain; Peroxisomal acyl-CoA oxidase 1, B chain; Peroxisomal acyl-CoA oxidase 1, C chain; Peroxisomal acyl-coenzyme A oxidase 1; peroxisomal fatty acyl-CoA oxidase; SCOX; Straight-chain acyl-CoA oxidase
Gene Aliases: ACOX; ACOX1; PALMCOX; SCOX
UniProt ID: (Human) Q15067
Entrez Gene ID: (Human) 51
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.