Proteintech
Manufacturer Code:665321IG150UL
Catalog # 50-173-6953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F10G5 |
Immunogen | ACOT9 Fusion Protein Ag8884 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACATE2 ACOT9 Acyl CoA thioester hydrolase 9 acyl CoA thioesterase 9 CGI 16 MT ACT48; Acyl-CoA thioester hydrolase 9; Acyl-CoA thioesterase 9; acyl-Coenzyme A thioesterase 2, mitochondrial; Acyl-coenzyme A thioesterase 9, mitochondrial; mitochondrial Acyl-CoA Thioesterase
Gene Aliases: ACATE2; ACOT9; CGI-16; MT-ACT48; MTACT48
UniProt ID: (Human) Q9Y305
Entrez Gene ID: (Human) 23597, (Rat) 302640
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.