Invitrogen
Manufacturer Code:PA5109356
Catalog # PIPA5109356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 22-250 of human ACOT9 (NP_001028755.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610041P13Rik Acate2 ACOT9 Acyl coenzyme A thioester hydrolase 2 Acyl-CoA thioester hydrolase 9 Acyl-CoA thioesterase 9 Acyl-coenzyme A thioesterase 9 mitochondrial C76421 CGI-16 Mitochondrial 48 kDa acyl-CoA thioester hydrolase 1 MTACT48 MT-ACT48 Mt-ACT48.1 MTE-2 p48 Protein U8 U8; Acyl coenzyme A thioester hydrolase 2; Acyl-CoA thioester hydrolase 9; Acyl-CoA thioesterase 9; acyl-Coenzyme A thioesterase 2, mitochondrial; Acyl-coenzyme A thioesterase 9, mitochondrial; Mitochondrial 48 kDa acyl-CoA thioester hydrolase 1; mitochondrial Acyl-CoA Thioesterase; Mt-ACT48.1; MTE-2; p48; Protein U8
Gene Aliases: 0610041P13Rik; ACATE2; ACOT9; C76421; CGI-16; MT-ACT48; MTACT48; p48; U8
UniProt ID: (Human) Q9Y305, (Mouse) Q9R0X4
Entrez Gene ID: (Human) 23597, (Mouse) 56360, (Rat) 302640
Molecular Function: esterase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.