Novus Biologicals
Manufacturer Code:NBP292103100UL
Catalog # NB026098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ACOT8 (NP_005460.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTEIII acyl-CoA thioesterase 8HNAACTE Choloyl-coenzyme A thioesterase EC 3.1.2.27 hACTEIII HIV-Nef-associated acyl-CoA thioesterase hTEcholoyl-CoA hydrolase long-chain fatty-acyl-CoA hydrolase palmitoyl-CoA hydrolase peroxisomal acyl-CoA thioesterase peroxisomal acyl-CoA thioesterase 1 Peroxisomal acyl-coenzyme A thioester hydrolase 1 Peroxisomal long-chain acyl-CoA thioesterase 1 PTE-1 PTE1HIV-Nef associated acyl-CoA thioesterase PTE2 PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8 Thioesterase II thioesterase III; Acyl-CoA thioesterase 8; Acyl-coenzyme A thioesterase 8; choloyl-CoA hydrolase; Choloyl-coenzyme A thioesterase; hACTE-III; HIV-Nef associated acyl-CoA thioesterase; HIV-Nef-associated acyl-CoA thioesterase; long-chain fatty-acyl-CoA hydrolase; Nef (lentivirus myristoylated factor) associated protein 1; palmitoyl-CoA hydrolase; peroxisomal acyl-CoA thioesterase 1; Peroxisomal acyl-CoA thioesterase 2; Peroxisomal acyl-coenzyme A thioester hydrolase 1; Peroxisomal long-chain acyl-CoA thioesterase 1; PTE-1; PTE-2; Thioesterase II; thioesterase III
Gene Aliases: ACOT8; ACTEIII; hACTE-III; HNAACTE; hTE; NAP1; PTE-1; PTE-2; PTE1; PTE2
UniProt ID: (Human) O14734
Entrez Gene ID: (Human) 10005
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.